Assay Details
| Gene Symbol: AspRS-m | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273590.1 | NP_001260519.1 | 5 - 6 | 1028 | 69 | |
| NM_001273591.1 | NP_001260520.2 | 4 - 5 | 1128 | 69 | ||
| NM_165196.2 | NP_724018.1 | 5 - 6 | 1162 | 69 | ||
| GenBank mRNA | AY119180.1 | - | 5 - 6 | 1142 | 69 | |
More Information
Set Membership: |
|

AspRS-m
Genomic Map