Assay Details
| Gene Symbol: Acsl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014507.2 | NP_001014507.1 | 3 - 4 | 614 | 56 | |
| NM_001014508.2 | NP_001014508.1 | 4 - 5 | 1148 | 56 | ||
| NM_001014509.2 | NP_001014509.1 | 3 - 4 | 795 | 56 | ||
| NM_001014510.2 | NP_001014510.1 | 3 - 4 | 670 | 56 | ||
| NM_001014511.2 | NP_001014511.1 | 5 - 6 | 882 | 56 | ||
| NM_001014512.2 | NP_001014512.1 | 3 - 4 | 713 | 56 | ||
| NM_001273881.1 | NP_001260810.1 | 4 - 5 | 774 | 56 | ||
| NM_143777.3 | NP_652034.2 | 4 - 5 | 938 | 56 | ||
| NM_170615.2 | NP_724695.1 | 5 - 6 | 965 | 56 | ||
| NM_170616.2 | NP_724696.1 | 3 - 4 | 830 | 56 | ||
| GenBank mRNA | AY094958.1 | - | 4 - 5 | 910 | 56 | |
More Information
Set Membership: |
|

Acsl
Genomic Map