Assay Details
| Gene Symbol: Acsl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014507.2 | NP_001014507.1 | 6 - 7 | 1472 | 61 | |
| NM_001014508.2 | NP_001014508.1 | 7 - 8 | 2006 | 61 | ||
| NM_001014509.2 | NP_001014509.1 | 6 - 7 | 1653 | 61 | ||
| NM_001014510.2 | NP_001014510.1 | 6 - 7 | 1528 | 61 | ||
| NM_001014511.2 | NP_001014511.1 | 8 - 9 | 1740 | 61 | ||
| NM_001014512.2 | NP_001014512.1 | 6 - 7 | 1571 | 61 | ||
| NM_001273881.1 | NP_001260810.1 | 7 - 8 | 1632 | 61 | ||
| NM_143777.3 | NP_652034.2 | 7 - 8 | 1796 | 61 | ||
| NM_170615.2 | NP_724695.1 | 8 - 9 | 1823 | 61 | ||
| NM_170616.2 | NP_724696.1 | 6 - 7 | 1688 | 61 | ||
| GenBank mRNA | AY094958.1 | - | 7 - 8 | 1768 | 61 | |
More Information
Set Membership: |
|

Acsl
Genomic Map