Assay Details
| Gene Symbol: shakB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103568.3 | NP_001097038.2 | 5 - 6 | 1918 | 133 | |
| NM_001144750.2 | NP_001138222.1 | 4 - 5 | 659 | 133 | ||
| NM_001258858.2 | NP_001245787.1 | 8 - 9 | 1770 | 133 | ||
| NM_001258859.2 | NP_001245788.1 | 9 - 10 | 2634 | 133 | ||
| NM_001298574.1 | NP_001285503.1 | 8 - 9 | 2875 | 133 | ||
| NM_078701.3 | NP_523425.2 | 7 - 8 | 2066 | 133 | ||
| NM_134566.3 | NP_608410.2 | 6 - 7 | 1170 | 133 | ||
| NM_170660.2 | NP_728361.1 | 4 - 5 | 1418 | 133 | ||
| GenBank mRNA | S78495.1 | - | 4 - 5 | 1418 | 133 | |
More Information
Set Membership: |
|

shakB
Genomic Map