Assay Details
| Gene Symbol: Plc21C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272889.1 | NP_001259818.1 | 2 - 3 | 832 | 65 | |
| NM_057503.5 | NP_476851.2 | 2 - 3 | 783 | 65 | ||
| NM_057504.5 | NP_476852.1 | 2 - 3 | 832 | 65 | ||
| NM_205882.2 | NP_995604.2 | 2 - 3 | 783 | 65 | ||
| NM_205883.2 | NP_995605.1 | 2 - 3 | 783 | 65 | ||
| NM_205884.3 | NP_995606.1 | 2 - 3 | 832 | 65 | ||
| GenBank mRNA | AY051657.1 | - | 2 - 3 | 795 | 65 | |
More Information
Set Membership: |
|

Plc21C
Genomic Map