Assay Details
| Gene Symbol: colt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057873.5 | NP_477221.1 | 1 | 1369 | 129 | |
| NM_205894.1 | NP_995616.1 | 1 | 1366 | 129 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

colt| Gene Symbol: colt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057873.5 | NP_477221.1 | 1 | 1369 | 129 | |
| NM_205894.1 | NP_995616.1 | 1 | 1366 | 129 | ||
Set Membership: |
|