Assay Details
| Gene Symbol: Syt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273036.1 | NP_001259965.1 | 6 - 7 | 2079 | 81 | |
| NM_001298638.1 | NP_001285567.1 | 9 - 10 | 1521 | 81 | ||
| NM_001298639.1 | NP_001285568.1 | 8 - 9 | 1518 | 81 | ||
| NM_001370000.1 | NP_001356906.1 | 9 - 10 | 2124 | 81 | ||
| NM_078736.3 | NP_523460.2 | 8 - 9 | 1524 | 81 | ||
| NM_164501.2 | NP_722838.1 | 8 - 9 | 1518 | 81 | ||
| NM_164502.2 | NP_722839.1 | 7 - 8 | 1167 | 81 | ||
| NM_205897.2 | NP_995619.2 | 7 - 8 | 1152 | 81 | ||
More Information
Set Membership: |
|

Syt1
Genomic Map