Assay Details
| Gene Symbol: eIF3h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080095.4 | NP_524834.2 | 1 - 2 | 439 | 101 | |
| NM_205909.3 | NP_995631.1 | 1 - 2 | 125 | 101 | ||
| GenBank mRNA | BT071794.1 | - | 1 - 2 | 81 | 101 | |
More Information
Set Membership: |
|

eIF3h
Genomic Map