Assay Details
| Gene Symbol: Ca-beta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259049.3 | NP_001245978.1 | 5 - 6 | 1439 | 84 | |
| NM_001259050.3 | NP_001245979.2 | 5 - 6 | 2128 | 84 | ||
| NM_001273446.1 | NP_001260375.1 | 5 - 6 | 2161 | 84 | ||
| NM_001273447.1 | NP_001260376.1 | 7 - 8 | 1489 | 84 | ||
| NM_001273448.2 | NP_001260377.1 | 5 - 6 | 2149 | 84 | ||
| NM_001298907.1 | NP_001285836.1 | 5 - 6 | 1430 | 84 | ||
| NM_078822.4 | NP_523546.1 | 5 - 6 | 1427 | 84 | ||
| NM_205962.3 | NP_995684.2 | 5 - 6 | 1427 | 84 | ||
| NM_205963.4 | NP_995685.3 | 5 - 6 | 2161 | 84 | ||
| GenBank mRNA | U11074.1 | - | 5 - 6 | 1025 | 84 | |
More Information
Set Membership: |
|

Ca-beta
Genomic Map