Assay Details
Gene Symbol: Vm26Ac | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_135129.2 | NP_608973.1 | 1 | 535 | 62 | |
GenBank mRNA | BT030874.1 | - | 1 | 533 | 62 | |
BT030882.1 | - | 1 | 533 | 62 | ||
BT031207.1 | - | 1 | 532 | 62 | ||
BT031223.1 | - | 1 | 535 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
