Assay Details
| Gene Symbol: Gal | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273191.1 | NP_001260120.1 | 3 - 4 | 567 | 77 | |
| NM_135134.3 | NP_608978.2 | 3 - 4 | 573 | 77 | ||
| GenBank mRNA | AY119076.1 | - | 3 - 4 | 537 | 77 | |
More Information
Set Membership: |
|

Gal
Genomic Map