Assay Details
| Gene Symbol: ND-51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298728.1 | NP_001285657.1 | 2 - 3 | 438 | 87 | |
| NM_135143.3 | NP_608987.1 | 2 - 3 | 475 | 87 | ||
| GenBank mRNA | AY051642.1 | - | 2 - 3 | 416 | 87 | |
More Information
Set Membership: |
|

ND-51
Genomic Map