Assay Details
| Gene Symbol: AANATL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298738.1 | NP_001285667.1 | 1 | 550 | 67 | |
| NM_135161.3 | NP_609005.1 | 1 | 550 | 67 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AANATL2| Gene Symbol: AANATL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298738.1 | NP_001285667.1 | 1 | 550 | 67 | |
| NM_135161.3 | NP_609005.1 | 1 | 550 | 67 | ||
Set Membership: |
|