Assay Details
| Gene Symbol: HemK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273205.2 | NP_001260134.1 | 1 - 2 | 301 | 60 | |
| NM_135187.4 | NP_609031.2 | 1 - 2 | 301 | 60 | ||
More Information
Set Membership: |
|

HemK1
Genomic Map