Assay Details
| Gene Symbol: Sec61alpha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298745.1 | NP_001285674.1 | 2 - 3 | 322 | 65 | |
| NM_135190.5 | NP_609034.1 | 2 - 3 | 213 | 65 | ||
| GenBank mRNA | AB062670.1 | - | 2 - 3 | 165 | 65 | |
| AY069569.1 | - | 2 - 3 | 182 | 65 | ||
More Information
Set Membership: |
|

Sec61alpha
Genomic Map