Assay Details
| Gene Symbol: CG11070 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135216.4 | NP_609060.1 | 1 - 2 | 802 | 86 | |
| GenBank mRNA | AF181657.1 | - | 1 - 2 | 775 | 86 | |
More Information
Set Membership: |
|

CG11070
Genomic Map