Assay Details
| Gene Symbol: Nse1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273216.1 | NP_001260145.1 | 1 | 745 | 65 | |
| NM_135219.3 | NP_609063.1 | 1 | 745 | 65 | ||
| GenBank mRNA | AY051646.1 | - | 1 | 726 | 65 | |
More Information
Set Membership: |
|

Nse1
Genomic Map