Assay Details
| Gene Symbol: Aatf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135222.3 | NP_609066.1 | 1 - 2 | 1452 | 68 | |
| GenBank mRNA | AY071275.1 | - | 1 - 2 | 1450 | 68 | |
More Information
Set Membership: |
|

Aatf
Genomic Map