Assay Details
| Gene Symbol: TTLL3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135225.2 | NP_609069.1 | 2 - 3 | 1461 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TTLL3A| Gene Symbol: TTLL3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135225.2 | NP_609069.1 | 2 - 3 | 1461 | 70 | |
Set Membership: |
|