Assay Details
| Gene Symbol: Gas41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135242.3 | NP_609086.1 | 1 - 2 | 610 | 65 | |
| GenBank mRNA | AY061214.1 | - | 1 - 2 | 538 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gas41| Gene Symbol: Gas41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135242.3 | NP_609086.1 | 1 - 2 | 610 | 65 | |
| GenBank mRNA | AY061214.1 | - | 1 - 2 | 538 | 65 | |
Set Membership: |
|