Assay Details
| Gene Symbol: Hmgcl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298767.1 | NP_001285696.1 | 1 - 2 | 112 | 57 | |
| NM_135245.3 | NP_609089.1 | 1 - 2 | 112 | 57 | ||
| GenBank mRNA | BT031041.1 | - | 1 - 2 | 63 | 57 | |
More Information
Set Membership: |
|

Hmgcl
Genomic Map