Assay Details
| Gene Symbol: MME1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135249.3 | NP_609093.1 | 3 - 4 | 1012 | 68 | |
| GenBank mRNA | AY069591.1 | - | 3 - 4 | 944 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MME1| Gene Symbol: MME1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135249.3 | NP_609093.1 | 3 - 4 | 1012 | 68 | |
| GenBank mRNA | AY069591.1 | - | 3 - 4 | 944 | 68 | |
Set Membership: |
|