Assay Details
| Gene Symbol: chm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259001.2 | NP_001245930.1 | 4 - 5 | 2044 | 70 | |
| NM_001273263.1 | NP_001260192.1 | 4 - 5 | 2044 | 70 | ||
| NM_135273.5 | NP_609117.4 | 4 - 5 | 2044 | 70 | ||
More Information
Set Membership: |
|

chm
Genomic Map