Assay Details
| Gene Symbol: SmE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135318.2 | NP_609162.1 | 1 | 447 | 83 | |
| GenBank mRNA | BT003597.1 | - | 1 | 422 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SmE| Gene Symbol: SmE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135318.2 | NP_609162.1 | 1 | 447 | 83 | |
| GenBank mRNA | BT003597.1 | - | 1 | 422 | 83 | |
Set Membership: |
|