Assay Details
| Gene Symbol: CG7367 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169435.1 | NP_001162906.1 | 3 - 4 | 536 | 93 | |
| NM_001259006.2 | NP_001245935.1 | 3 - 4 | 497 | 93 | ||
| NM_135331.2 | NP_609175.2 | 2 - 3 | 308 | 93 | ||
| GenBank mRNA | BT022244.1 | - | 3 - 4 | 536 | 93 | |
| BT022296.1 | - | 3 - 4 | 473 | 93 | ||
More Information
Set Membership: |
|

CG7367
Genomic Map