Assay Details
| Gene Symbol: RpL36A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298808.1 | NP_001285737.1 | 2 - 3 | 265 | 63 | |
| NM_135335.4 | NP_609179.2 | 2 - 3 | 215 | 63 | ||
| GenBank mRNA | BT029953.1 | - | 1629 | 63 | ||
More Information
Set Membership: |
|

RpL36A
Genomic Map