Assay Details
| Gene Symbol: PAPLA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298812.1 | NP_001285741.1 | 1 - 2 | 416 | 63 | |
| NM_135341.1 | NP_609185.1 | 1 - 2 | 353 | 63 | ||
| GenBank mRNA | BT023778.1 | - | 1 - 2 | 353 | 63 | |
More Information
Set Membership: |
|

PAPLA1
Genomic Map