Assay Details
| Gene Symbol: PAPLA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201785.1 | NP_001188714.1 | 5 - 6 | 5141 | 83 | |
| NM_001201786.2 | NP_001188715.1 | 5 - 6 | 5044 | 83 | ||
| NM_135341.1 | NP_609185.1 | 5 - 6 | 5268 | 83 | ||
| GenBank mRNA | BT023778.1 | - | 5 - 6 | 5268 | 83 | |
More Information
Set Membership: |
|

PAPLA1
Genomic Map