Assay Details
| Gene Symbol: Rbsn-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135342.5 | NP_609186.1 | 2 - 3 | 1544 | 96 | |
| GenBank mRNA | AY061374.1 | - | 2 - 3 | 1515 | 96 | |
More Information
Set Membership: |
|

Rbsn-5
Genomic Map