Assay Details
| Gene Symbol: Khc-73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274070.2 | NP_001260999.1 | 5 - 6 | 1488 | 62 | |
| NM_001274071.2 | NP_001261000.1 | 6 - 7 | 1749 | 62 | ||
| NM_001299533.1 | NP_001286462.1 | 6 - 7 | 1749 | 62 | ||
| NM_135357.5 | NP_609201.3 | 6 - 7 | 1749 | 62 | ||
| NM_176176.3 | NP_788356.2 | 6 - 7 | 1749 | 62 | ||
More Information
Set Membership: |
|

Khc-73
Genomic Map