Assay Details
| Gene Symbol: wol | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135358.2 | NP_609202.1 | 2 - 3 | 405 | 78 | |
| GenBank mRNA | AY094692.1 | - | 2 - 3 | 390 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

wol| Gene Symbol: wol | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135358.2 | NP_609202.1 | 2 - 3 | 405 | 78 | |
| GenBank mRNA | AY094692.1 | - | 2 - 3 | 390 | 78 | |
Set Membership: |
|