Assay Details
| Gene Symbol: Rcd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201793.1 | NP_001188722.1 | 1 - 2 | 355 | 122 | |
| NM_135377.3 | NP_609221.1 | 1 - 2 | 407 | 122 | ||
| GenBank mRNA | AY119223.1 | - | 1 - 2 | 355 | 122 | |
More Information
Set Membership: |
|

Rcd4
Genomic Map