Assay Details
| Gene Symbol: Dad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135378.4 | NP_609222.1 | 1 | 444 | 84 | |
| GenBank mRNA | AY113429.1 | - | 1 | 440 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dad1| Gene Symbol: Dad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135378.4 | NP_609222.1 | 1 | 444 | 84 | |
| GenBank mRNA | AY113429.1 | - | 1 | 440 | 84 | |
Set Membership: |
|