Assay Details
| Gene Symbol: Bace | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298827.1 | NP_001285756.1 | 1 | 1137 | 89 | |
| NM_135391.3 | NP_609235.1 | 1 | 1137 | 89 | ||
| GenBank mRNA | BT016133.1 | - | 1 | 1119 | 89 | |
More Information
Set Membership: |
|

Bace
Genomic Map