Assay Details
| Gene Symbol: PIG-U | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135394.3 | NP_609238.1 | 1 - 2 | 184 | 74 | |
| GenBank mRNA | AY058668.1 | - | 1 - 2 | 164 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PIG-U| Gene Symbol: PIG-U | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135394.3 | NP_609238.1 | 1 - 2 | 184 | 74 | |
| GenBank mRNA | AY058668.1 | - | 1 - 2 | 164 | 74 | |
Set Membership: |
|