Assay Details
| Gene Symbol: CG9541 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273344.1 | NP_001260273.1 | 2 - 3 | 382 | 80 | |
| NM_135419.3 | NP_609263.2 | 2 - 3 | 834 | 80 | ||
More Information
Set Membership: |
|

CG9541
Genomic Map