Assay Details
| Gene Symbol: CG9541 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135419.3 | NP_609263.2 | 5 - 6 | 1264 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CG9541| Gene Symbol: CG9541 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135419.3 | NP_609263.2 | 5 - 6 | 1264 | 68 | |
Set Membership: |
|