Assay Details
| Gene Symbol: CG9541 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273344.1 | NP_001260273.1 | 5 - 6 | 1210 | 77 | |
| NM_135419.3 | NP_609263.2 | 6 - 7 | 1572 | 77 | ||
More Information
Set Membership: |
|

CG9541
Genomic Map