Assay Details
Gene Symbol: Npc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201840.2 | NP_001188769.1 | 11 - 12 | 3956 | 80 | |
NM_001201841.2 | NP_001188770.1 | 11 - 12 | 3918 | 80 | ||
NM_001201842.2 | NP_001188771.1 | 11 - 12 | 3884 | 80 | ||
NM_001298866.1 | NP_001285795.1 | 11 - 12 | 4048 | 80 | ||
NM_001298867.1 | NP_001285796.1 | 11 - 12 | 3869 | 80 | ||
NM_001298868.1 | NP_001285797.1 | 11 - 12 | 4862 | 80 | ||
NM_135513.5 | NP_609357.2 | 11 - 12 | 4060 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
