Assay Details
| Gene Symbol: Mob3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135520.4 | NP_609364.1 | 5 - 6 | 720 | 116 | |
| GenBank mRNA | AY071000.1 | - | 5 - 6 | 719 | 116 | |
More Information
Set Membership: |
|

Mob3
Genomic Map