Assay Details
| Gene Symbol: CG4968 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135531.3 | NP_609375.1 | 1 - 2 | 200 | 93 | |
| GenBank mRNA | AY061382.1 | - | 1 - 2 | 217 | 93 | |
More Information
Set Membership: |
|

CG4968
Genomic Map