Assay Details
| Gene Symbol: CG4968 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135531.3 | NP_609375.1 | 2 - 3 | 500 | 78 | |
| GenBank mRNA | AY061382.1 | - | 2 - 3 | 517 | 78 | |
More Information
Set Membership: |
|

CG4968
Genomic Map