Assay Details
| Gene Symbol: ClpP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273403.1 | NP_001260332.1 | 1 - 2 | 165 | 90 | |
| NM_135544.3 | NP_609388.1 | 1 - 2 | 165 | 90 | ||
| GenBank mRNA | AY119497.1 | - | 1 - 2 | 176 | 90 | |
More Information
Set Membership: |
|

ClpP
Genomic Map