Assay Details
| Gene Symbol: gny | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273409.1 | NP_001260338.1 | 1 - 2 | 711 | 71 | |
| NM_135549.4 | NP_609393.1 | 1 - 2 | 711 | 71 | ||
| GenBank mRNA | AY051761.1 | - | 1 - 2 | 696 | 71 | |
More Information
Set Membership: |
|

gny
Genomic Map