Assay Details
| Gene Symbol: CG5188 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135557.4 | NP_609401.2 | 2 - 3 | 780 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CG5188| Gene Symbol: CG5188 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135557.4 | NP_609401.2 | 2 - 3 | 780 | 86 | |
Set Membership: |
|