Assay Details
| Gene Symbol: CG7309 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135624.5 | NP_609468.4 | 8 - 9 | 1584 | 89 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CG7309| Gene Symbol: CG7309 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135624.5 | NP_609468.4 | 8 - 9 | 1584 | 89 | |
Set Membership: |
|