Assay Details
| Gene Symbol: Reps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259046.2 | NP_001245975.1 | 2 - 3 | 983 | 101 | |
| NM_135643.4 | NP_609487.2 | 2 - 3 | 983 | 101 | ||
| GenBank mRNA | AY128416.1 | - | 2 - 3 | 979 | 101 | |
More Information
Set Membership: |
|

Reps
Genomic Map