Assay Details
| Gene Symbol: l(2)gd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259047.2 | NP_001245976.1 | 2 - 3 | 1780 | 85 | |
| NM_135644.4 | NP_609488.1 | 2 - 3 | 1780 | 85 | ||
| GenBank mRNA | AY058523.1 | - | 2 - 3 | 1755 | 85 | |
More Information
Set Membership: |
|

l(2)gd1
Genomic Map