Assay Details
| Gene Symbol: dl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169529.1 | NP_001163000.1 | 8 - 9 | 2096 | 115 | |
| NM_001169530.1 | NP_001163001.1 | 8 - 9 | 2010 | 115 | ||
| NM_165217.3 | NP_724052.1 | 9 - 10 | 1995 | 115 | ||
| NM_165218.3 | NP_724053.1 | 9 - 10 | 2339 | 115 | ||
| GenBank mRNA | BT099643.1 | - | 5 - 6 | 5701 | 115 | |
| BT133461.1 | - | 9 - 10 | 1991 | 115 | ||
More Information
Set Membership: |
|

dl
Genomic Map