Assay Details
| Gene Symbol: BicD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259128.2 | NP_001246057.1 | 4 - 5 | 2475 | 84 | |
| NM_001273601.1 | NP_001260530.1 | 4 - 5 | 2475 | 84 | ||
| NM_001273602.1 | NP_001260531.1 | 4 - 5 | 2475 | 84 | ||
| NM_165220.3 | NP_724056.1 | 4 - 5 | 2475 | 84 | ||
| GenBank mRNA | AY069452.1 | - | 4 - 5 | 1978 | 84 | |
| X51652.1 | - | 2518 | 84 | |||
More Information
Set Membership: |
|

BicD
Genomic Map